Skip to Content
Merck
All Photos(2)

Key Documents

SRP5285

Sigma-Aldrich

NLK, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp761G1211, FLJ21033

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
3.570,00 kr.

3.570,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
3.570,00 kr.

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

3.570,00 kr.


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1.9-2.9 nmol/min·mg

mol wt

~84 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NLK(51701)

General description

NLK (also known as nemo-like kinase) is a member of the serine/threonine protein kinase that is encoded by the NLK gene. NLK plays a role in cell fate determination and is required for the differentiation of bone marrow stromal cells. NLK also suppresses a wide range of transcription factors such as CREB by phosphorylation of the co-activator CREBBP. NLK expression increases the number of apoptotic cells but does not induce obvious cell cycle arrest. NLK suppresses Notch signalling by interfering with formation of the Notch active transcriptional complex.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service