Skip to Content
Merck
All Photos(1)

Documents

SRP5151

Sigma-Aldrich

XIAP, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

API3, BIRC4, ILP1, MIHA, XLP2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~84 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... XIAP(331)

General description

XIAP is a member of a family of proteins which inhibit apoptosis and act as mammalian cell-death suppressors. XIAP inhibits at least 2 members of the caspase family of cell-death proteases, caspase-3 and caspase-7. In addition, XIAP inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. Furthermore, XIAP can mediate protection of cells from apoptosis by utilizing both a JNK1 activation pathway that involves ILPIP and a caspase inhibition pathway that is independent of ILPIP. Disruption of the XIAP gene in human colon cancer cells can enhance sensitivity of these cells to exogenously added TRAIL suggesting that XIAP is a nonredundant modulator of TRAIL-mediated apoptosis.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Tang et al.
Nature, 414(6861), 313-317 (2001-11-20)
The proinflammatory cytokine tumour necrosis factor-alpha (TNF-alpha) regulates immune responses, inflammation and programmed cell death (apoptosis). The ultimate fate of a cell exposed to TNF-alpha is determined by signal integration between its different effectors, including IkappaB kinase (IKK), c-Jun N-terminal
Q L Deveraux et al.
Nature, 388(6639), 300-304 (1997-07-17)
The inhibitor-of-apoptosis (IAP) family of genes has an evolutionarily conserved role in regulating programmed cell death in animals ranging from insects to humans. Ectopic expression of human IAP proteins can suppress cell death induced by a variety of stimuli, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service