Skip to Content
Merck
All Photos(1)

Key Documents

SRP0704

Sigma-Aldrich

BML-259

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
2.380,00 kr.

2.380,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
25 MG
2.380,00 kr.

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

2.380,00 kr.


Please contact Customer Service for Availability

Assay

>95% (TLC)

form

lyophilized powder

mol wt

~260.4

shipped in

ambient

storage temp.

−20°C

Biochem/physiol Actions

Potent inhibitor of CDK5/p25 (IC50=64 nM). Inhibits CDK2/cyclin E at slightly higher concentration (IC50=98nM) (1). Protects cells against NAC-amyloid-induced cell death (2). Cell permeable. MW=260.4. Purity ?95% by TLC. Formulated as a lyophilized powder. May be dissolved in DMSO (>25 mg/mL) or DMF (30 mg/mL). Stable desicated as supplied at ambient temperature for up to 2 years, or in solution at -20°C for up to 2 months.

Storage and Stability

Store desiccated as supplied at -20°C for up to 2 years. Store solutions at -20°C for up to 2 months.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service