Skip to Content
Merck
All Photos(1)

Key Documents

SRP0414

Sigma-Aldrich

PKLR Var2 active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

PK1, PKL, RPK, pyruvate kinase, liver and RBC

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
4.440,00 kr.

4.440,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
4.440,00 kr.

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

4.440,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

59 kDa

packaging

pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PKLR(5313)

General description

PKR (GenBank Accession No. NM_181871) full length, amino acids 2-543 (end) with N-terminal His-tag, MW=59 kDa, expressed in an E. coli expression system.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven T Russell et al.
Experimental cell research, 315(1), 16-25 (2008-11-01)
Treatment of murine myotubes with high glucose concentrations (10 and 25 mM) stimulated protein degradation through the ubiquitin-proteasome pathway, and also caused activation (autophosphorylation) of PKR (double-stranded-RNA-dependent protein kinase) and eIF2alpha (eukaryotic initiation factor 2alpha). Phosphorylation of PKR and eIF2alpha
María J Bullido et al.
Neurobiology of aging, 29(8), 1160-1166 (2007-04-11)
Sporadic Alzheimer's disease (AD) appears to be the consequence of the interaction between combinations of genes and environmental factors (for example virus infections). To test this hypothesis, we are examining human genes relevant to herpes simplex virus type 1 (HSV-1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service