Skip to Content
Merck
All Photos(2)

Documents

SML3803

Sigma-Aldrich

Gabapentin Enacarbil

≥95% (HPLC)

Synonym(s):

1-[[(1-(Isobutanoyloxy)ethoxy)carbonyl]aminomethyl]cyclohex-1-ylacetic acid, 1-[[[[1-(2-Methyl-1-oxopropoxy)ethoxy]carbonyl]amino]methyl]cyclohexaneacetic acid, XP 13512

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27NO6
CAS Number:
Molecular Weight:
329.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥95% (HPLC)

form

oil

color

, Faint yellow to dark yellow

storage temp.

2-8°C

Biochem/physiol Actions

Gabapentin enacarbil, a prodrug to gabapentin, is an orally available anticonvulsant and analgesic drug of the gabapentinoid class. Gabapentin enacarbil is used for treatment of restless legs syndrome.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

XP13512 [(+/-)-1-([(alpha-isobutanoyloxyethoxy)carbonyl] aminomethyl)-1-cyclohexane acetic acid], a novel gabapentin prodrug: II. Improved oral bioavailability, dose proportionality, and colonic absorption compared with gabapentin in rats and monkeys
Journal of Pharmacology and Experimental Therapeutics, 311(1), 324-333 (2004)
Non-dopaminergic vs. dopaminergic treatment options in restless legs syndrome
Advances in Pharmacology, 84, 187-205 (2019)
Satomi Yamaguchi Ikeuchi et al.
Biological & pharmaceutical bulletin, 41(11), 1708-1715 (2018-11-02)
The purpose of this research was to establish an in vitro dissolution testing method to predict the oral pharmacokinetic (PK) profiles and food effects of gabapentin enacarbil formulated as wax matrix extended-release (ER) tablets in humans. We adopted various biorelevant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service