Skip to Content
Merck
All Photos(2)

Key Documents

SML3714

Sigma-Aldrich

SMIP004-7

≥98% (HPLC)

Synonym(s):

2-Methyl-4-butylaniline, 4-Butyl-2-methylaniline, 4-Butyl-2-methylbenzenamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
1.370,00 kr.
25 MG
5.510,00 kr.

1.370,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
1.370,00 kr.
25 MG
5.510,00 kr.

About This Item

Empirical Formula (Hill Notation):
C11H17N
CAS Number:
Molecular Weight:
163.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

1.370,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

(Oil or liquid)

color

colorless to brown

storage temp.

-10 to -25°C

SMILES string

Nc1c(cc(cc1)CCCC)C

InChI key

JTXOXRXZCAMPHL-UHFFFAOYSA-N

Biochem/physiol Actions

SMIP004-7 is a cell penetrant, potent and selective ubiquinone uncompetitive inhibitor of mitochondrial Complex I (NADH:ubiquinone oxidoreductase). SMIP004-7 selectively binds to a distinct N-terminal region of Complex I catalytic subunit NDUFS2, which leads to rapid disassembly of Complex I. SMIP004-7 potently inhibits growth of triple-negative breast cancer xenografts in mice.
cell penetrant, potent and selective ubiquinone uncompetitive inhibitor of mitochondrial Complex I

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic targeting of cancer by a ubiquinone uncompetitive inhibitor of mitochondrial complex I
Cell Chemical Biology, 29(3), 436-450 (2022)
Elizabeth Rico-Bautista et al.
Oncotarget, 4(8), 1212-1229 (2013-08-02)
We previously identified SMIP004 (N-(4-butyl-2-methyl-phenyl) acetamide) as a novel inducer of cancer-cell selective apoptosis of human prostate cancer cells. SMIP004 decreased the levels of positive cell cycle regulators, upregulated cyclin-dependent kinase inhibitors, and resulted in G1 arrest, inhibition of colony
Gabriela Reyes-Castellanos et al.
Trends in pharmacological sciences, 43(9), 706-708 (2022-04-07)
Targeting metabolic reprogramming has proven successful in oncology, but this field requires better identification of drugs that inhibit mitochondrial metabolism in cancer cells. Recent work from Dr Wolf's group reveals that the primary target of the antitumor compound SMIP004-7 is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service