Skip to Content
Merck
All Photos(1)

Key Documents

SML3406

Sigma-Aldrich

Nutlin-3b

≥98% (HPLC)

Synonym(s):

4-[[(4R,5S)-4,5-Bis(4-chlorophenyl)-4,5-dihydro-2-[4-methoxy-2-(1-methylethoxy)phenyl]-1H-imidazol-1-yl]carbonyl]-2-piperazinone, Nutlin 3b

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
779,00 kr.
25 MG
3.130,00 kr.

779,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
779,00 kr.
25 MG
3.130,00 kr.

About This Item

Empirical Formula (Hill Notation):
C30H30Cl2N4O4
CAS Number:
Molecular Weight:
581.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

779,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC(C=C1)=CC=C1[C@@H]2[C@H](C3=CC=C(Cl)C=C3)N(C(N4CCNC(C4)=O)=O)C(C5=C(OC(C)C)C=C(OC)C=C5)=N2

InChI

1S/C30H30Cl2N4O4/c1-18(2)40-25-16-23(39-3)12-13-24(25)29-34-27(19-4-8-21(31)9-5-19)28(20-6-10-22(32)11-7-20)36(29)30(38)35-15-14-33-26(37)17-35/h4-13,16,18,27-28H,14-15,17H2,1-3H3,(H,33,37)/t27-,28+/m1/s1

InChI key

BDUHCSBCVGXTJM-IZLXSDGUSA-N

Biochem/physiol Actions

Nutlin-3b, a (+)-enantiomer of Nutlin-3, is a weakly potent antagonist of p53/MDM2 (IC50 = 13.6 μM) that can serve as negative control for Nutlin-3a (IC50 = 90 nM) and other Mdm2 inhibitors.
weakly potent antagonist of p53/MDM2 that can serve as negative control for Nutlin-3a

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service