Skip to Content
Merck
All Photos(1)

Key Documents

SML3351

Sigma-Aldrich

Br-DAPI hydrochloride

≥95% (HPLC)

Synonym(s):

2-[4-(Aminoiminomethyl)phenyl]-3-bromo-1H-indole-6-carboximidamide hydrochloride, 3-Bromo-2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
834,00 kr.
25 MG
3.360,00 kr.

834,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
834,00 kr.
25 MG
3.360,00 kr.

About This Item

Empirical Formula (Hill Notation):
C16H14BrN5 · xHCl
Molecular Weight:
356.22 (free base basis)
UNSPSC Code:
41116158
NACRES:
NA.77

834,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

BrC1=C(NC2=CC(C(N)=N)=CC=C12)C3=CC=C(C=C3)C(N)=N

InChI

1S/C16H14BrN5/c17-13-11-6-5-10(16(20)21)7-12(11)22-14(13)8-1-3-9(4-2-8)15(18)19/h1-7,22H,(H3,18,19)(H3,20,21)

InChI key

GSBXNZFEZFIMCS-UHFFFAOYSA-N

Biochem/physiol Actions

Br-DAPI is a DAPI-derived DNA-binding photosensitizer that produces reactive oxygen species (ROS) upon irradiation, resulting in single/double stranded breaks of bound DNA. Light-induced viability reduction is observed among A549 cells treated with Br-DAPI (IC50 = 1.3 μM; 45 min treatment), but not DAPI (IC50 >64 μM) upon photosensitization (365 nm 15 mW/cm2, 4.5 J/cm2 fluence rate). Its high permeation in both Gram-negative and Gram-positive bacteria allows selective light-induced eradication of bacteria over mammalian cells at low concentration (IC50 = 0.29 μM/N99 E. coli and 0.5 μM/B. subtilis) and light intensity (5 min irradiation by 365 nm LED at 15 mW/cm2, 4.5 J/cm2).
DAPI-derived DNA-binding photosensitizer that allows light-induced cancer cell killing at high concetrations and selective bacteria eradication at low concentrations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service