Skip to Content
Merck
All Photos(1)

Key Documents

SML3040

Sigma-Aldrich

ZnDA-1H

≥98% (HPLC)

Synonym(s):

4-(3-(Bis(pyridin-2-ylmethyl)amino)-2-oxo-7,8-dihydro-2H-pyrano[3,2-g]quinolin-9(6H)-yl)-N-(2-(2-((6-chlorohexyl)oxy)ethoxy)ethyl)butanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
926,00 kr.
25 MG
3.750,00 kr.

926,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
926,00 kr.
25 MG
3.750,00 kr.

About This Item

Empirical Formula (Hill Notation):
C38H48ClN5O5
Molecular Weight:
690.27
UNSPSC Code:
12352200
NACRES:
NA.77

926,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ZnDA-1H is a highly selective fluorescent probe for Zn2+ that exhibits a reversible fluorescence response toward labile Zn2+ in various intracellular organelles, including the nucleus, mitochondria, endoplasmic reticulum, and Golgi apparatus in live cells. ZnDA-1H is not susceptible to intracellular pH variation
highly selective fluorescent probe for Zn2+ that exhibits a reversible fluorescence response toward labile Zn2+ in various intracellular organelles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiyuki Kowada et al.
Cell chemical biology, 27(12), 1521-1531 (2020-10-01)
Fluorescent Zn2+ probes used for the quantitative analysis of labile Zn2+ concentration ([Zn2+]) in target organelles are crucial for understanding the role of Zn2+ in biological processes. Although several fluorescent Zn2+ probes have been developed to date, there is still

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service