Skip to Content
Merck
All Photos(1)

Key Documents

SML2818

Sigma-Aldrich

BAY-207

≥98% (HPLC)

Synonym(s):

N-(4-(3-Chlorophenoxy)-3-sulfamoylphenyl)-1-(2-chlorophenyl)cyclopropanecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
514,00 kr.
25 MG
2.070,00 kr.

514,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
514,00 kr.
25 MG
2.070,00 kr.

About This Item

Empirical Formula (Hill Notation):
C22H18Cl2N2O4S
CAS Number:
Molecular Weight:
477.36
UNSPSC Code:
12352200
NACRES:
NA.77

514,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N)c1c(ccc(c1)NC(=O)C4(CC4)c3c(cccc3)Cl)Oc2cc(ccc2)Cl

InChI

1S/C22H18Cl2N2O4S/c23-14-4-3-5-16(12-14)30-19-9-8-15(13-20(19)31(25,28)29)26-21(27)22(10-11-22)17-6-1-2-7-18(17)24/h1-9,12-13H,10-11H2,(H,26,27)(H2,25,28,29)

InChI key

JXAOITUGCKTSTF-UHFFFAOYSA-N

Biochem/physiol Actions

BAY-207 is a negative control for a potent and selective P2X4 antagonist BAY-1797.
negative control for a potent and selective P2X4 antagonist BAY-1797

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Werner et al.
Journal of medicinal chemistry, 62(24), 11194-11217 (2019-11-21)
The P2X4 receptor is a ligand-gated ion channel that is expressed on a variety of cell types, especially those involved in inflammatory and immune processes. High-throughput screening led to a new class of P2X4 inhibitors with substantial CYP 3A4 induction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service