Skip to Content
Merck
All Photos(1)

Key Documents

SML2616

Sigma-Aldrich

Bepotastine besilate

≥98% (HPLC)

Synonym(s):

(+)-4-[[(S)-p-chloro-alpha-2­pyridylbenzyl]oxy]-1-piperidine butyric acid monobenzenesulfonate, Bepreve

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
500,00 kr.
50 MG
2.040,00 kr.

500,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
500,00 kr.
50 MG
2.040,00 kr.

About This Item

Empirical Formula (Hill Notation):
C21H25ClN2O3 · C6H6O3S
CAS Number:
Molecular Weight:
547.06
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

500,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 4.5 to 6.5°, c = 1 in methanol

storage condition

desiccated

color

white to beige

storage temp.

−20°C

SMILES string

[S](=O)(=O)(O)c4ccccc4.Clc1ccc(cc1)[C@@H](OC3CCN(CC3)CCCC(=O)O)c2ncccc2

InChI

1S/C21H25ClN2O3.C6H6O3S/c22-17-8-6-16(7-9-17)21(19-4-1-2-12-23-19)27-18-10-14-24(15-11-18)13-3-5-20(25)26;7-10(8,9)6-4-2-1-3-5-6/h1-2,4,6-9,12,18,21H,3,5,10-11,13-15H2,(H,25,26);1-5H,(H,7,8,9)/t21-;/m1./s1

InChI key

UDGHXQPQKQPSBB-ZMBIFBSDSA-N

Biochem/physiol Actions

Bepotastine besilate is a second generation antihistamine used clinically for the treatment of itching associated with allergic conjunctivitis, allergic rhinitis and urticaria/pruritus. It is a direct H1-receptor antagonist that inhibits the release of histamine from mast cells.
Second generation antihistamine, H1-receptor antagonist

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service