Skip to Content
Merck
All Photos(1)

Key Documents

SML2586

Sigma-Aldrich

(S)-C33

≥98% (HPLC)

Synonym(s):

6-[[(1S)-1-(4-Chlorophenyl)ethyl]amino]-1-cyclopentyl-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one, C33(S)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20ClN5O
CAS Number:
Molecular Weight:
357.84
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2=C(N=C(N1)N[C@H](C3=CC=C(C=C3)Cl)C)N(C4CCCC4)N=C2

Biochem/physiol Actions

(S)-C33 [C33(S)] is a potent and selective phosphodiesterase-9A (PDE9A) inhibitor that upregulates cGMP signaling. (S)-C33 protects against cardiac hypertrophy and postpones the transition to heart failure in rat models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manna Huang et al.
Molecular pharmacology, 88(5), 836-845 (2015-09-01)
Phosphodiesterase-9 (PDE9) inhibitors have been studied as potential therapeutics for treatment of central nervous system diseases and diabetes. Here, we report the discovery of a new category of PDE9 inhibitors by rational design on the basis of the crystal structures.
Pan-Xia Wang et al.
Acta pharmacologica Sinica, 38(9), 1257-1268 (2017-06-27)
Phosphodiesterase-9A (PDE9A) expression is upregulated during cardiac hypertrophy and heart failure. Accumulating evidence suggests that PDE9A might be a promising therapeutic target for heart diseases. The present study sought to investigate the effects and underlying mechanisms of C33(S), a novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service