Skip to Content
Merck
All Photos(1)

Key Documents

SML2349

Sigma-Aldrich

BAY-604

≥97% (HPLC)

Synonym(s):

(S)-N-Ethyl-1-methyl-4-(3-methylmorpholino)-1H-pyrrolo[2,3-b]pyridine-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
773,00 kr.
25 MG
3.000,00 kr.

773,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
773,00 kr.
25 MG
3.000,00 kr.

About This Item

Empirical Formula (Hill Notation):
C16H22N4O2
CAS Number:
Molecular Weight:
302.37
UNSPSC Code:
31401503
NACRES:
NA.77

773,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

color

white to very dark brown

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCC)C1=CC2=C(N1C)N=CC=C2N3CCOC[C@@H]3C

InChI key

SYYSMZCCSNLTHZ-NSHDSACASA-N

Biochem/physiol Actions

BAY-604 is a control probe for BAY-707, an inhibitor of the human 7,8-dihydro-8-oxoguaninetriphosphatase MTH1 (NUDT1)
Control probe for BAY-707, an inhibitor of the human 7,8-dihydro-8-oxoguaninetriphosphatase MTH1 (NUDT1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fredrik Rahm et al.
Journal of medicinal chemistry, 61(6), 2533-2551 (2018-02-28)
Recent literature has both suggested and questioned MTH1 as a novel cancer target. BAY-707 was just published as a target validation small molecule probe for assessing the effects of pharmacological inhibition of MTH1 on tumor cell survival, both in vitro

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service