Skip to Content
Merck
All Photos(1)

Key Documents

SML1873

Sigma-Aldrich

(2R,6R)-Hydroxynorketamine hydrochloride

≥98% (HPLC)

Synonym(s):

(2R,6R)-HNK hydrochloride; (2R,6R)-2-Amino-2-(2-chlorophenyl)-6-hydroxy-cyclohexanone hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
1.070,00 kr.
25 MG
4.330,00 kr.

1.070,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
1.070,00 kr.
25 MG
4.330,00 kr.

About This Item

Empirical Formula (Hill Notation):
C12H14ClNO2 · HCl
CAS Number:
Molecular Weight:
276.16
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.77

1.070,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -95 to -115°, c = 1 in H2O

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

desiccated

color

white to beige

solubility

H2O: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1[C@@](C2=CC=CC=C2Cl)(N)CCC[C@H]1O.Cl

InChI

1S/C12H14ClNO2.ClH/c13-9-5-2-1-4-8(9)12(14)7-3-6-10(15)11(12)16;/h1-2,4-5,10,15H,3,6-7,14H2;1H/t10-,12-;/m1./s1

InChI key

ZQJVHBJDLMIKJK-MHDYBILJSA-N

Biochem/physiol Actions

(2R,6R)-Hydroxynorketamine is a ketamine metabolite that has rapid antidepressant activity without ketamine-related side effects.
(2R,6R)-Hydroxynorketamine is a ketamine metabolite that has rapid antidepressant activity without ketamine-related side effects. The NMDAR antagonist (R,S)-ketamine must be metabolized to (2S,6S;2R,6R)-hydroxynorketamine (HNK) to have antidepressant effects. The (2R,6R)-HNK enantiomer appears to be the enantiomer most responsible for antidepressant effects in a mouse study and unlike (2S,6S)-HNK, (2R,6R)-HNK caused no significant changes in locomotion or coordination. (2R,6R)-HNK does not seem to have the same addiction potential as ketamine, since mice did not self-administer pharmacologically relevant doses of (2R,6R)-HNK under the same conditions where ketamine was readily self-administered. The (2R,6R)-HNK antidepressant appears to involve activation of AMPA glutamate receptors rather than inhibition of NMDA glutamate receptors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Panos Zanos et al.
Nature, 533(7604), 481-486 (2016-05-05)
Major depressive disorder affects around 16 per cent of the world population at some point in their lives. Despite the availability of numerous monoaminergic-based antidepressants, most patients require several weeks, if not months, to respond to these treatments, and many

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service