Skip to Content
Merck
All Photos(1)

Documents

SML1521

Sigma-Aldrich

Irsogladine maleate

≥98% (HPLC)

Synonym(s):

2,4-Diamino-6-(2,5-dichlorophenyl)-s-triazine maleate, 6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine (2Z)-2-butenedioate (1:1), Gaslon N, MN 1695

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7Cl2N5 ·C4H4O4
CAS Number:
Molecular Weight:
372.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

room temp

SMILES string

C[C@]1([H])[C@@](C)(O)CC2=C(C(OC)=C(OC)C(OC)=C2)C3=C(OC)C4=C(OCO4)C=C3C1

InChI

1S/C23H28O7/c1-12-7-13-8-16-20(30-11-29-16)22(28-6)17(13)18-14(10-23(12,2)24)9-15(25-3)19(26-4)21(18)27-5/h8-9,12,24H,7,10-11H2,1-6H3/t12-,23-/m0/s1

InChI key

ZWRRJEICIPUPHZ-MYODQAERSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Irsogladine maleate is an inhibitor of phosphodiesterase 4 (PDE4) with gastroprotective activity. Irsogladine maleate is a mucosal protective drug with a number of activites including facilitation of gap junctional intercellular communication, inhibition of the reduced gastric mucosal blood flow response, and suppression of reactive oxygen species (ROS) production.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service