Skip to Content
Merck
All Photos(1)

Key Documents

SML1245

Sigma-Aldrich

ML396

≥98% (HPLC)

Synonym(s):

N-(3-Chloro-4-((5-chloropyridin-2-yl)oxy)phenyl)picolinamide, VU0422288

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
581,00 kr.

581,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
581,00 kr.

About This Item

Empirical Formula (Hill Notation):
C17H11Cl2N3O2
CAS Number:
Molecular Weight:
360.19
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

581,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, white to yellow to brown

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(OC2=NC=C(Cl)C=C2)C(Cl)=C1)C3=NC=CC=C3

InChI

1S/C17H11Cl2N3O2/c18-11-4-7-16(21-10-11)24-15-6-5-12(9-13(15)19)22-17(23)14-3-1-2-8-20-14/h1-10H,(H,22,23)

InChI key

MZRLPXFGQKQHST-UHFFFAOYSA-N

Biochem/physiol Actions

ML396 is a potent positive allosteric modulator or metabotropic glutamate receptors group III (mGlu group III PAM).
ML396 is a potent positive allosteric modulator or metabotropic glutamate receptors group III (mGlu group III PAM). ML396 was used to study the biology of mGlu7.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service