Skip to Content
Merck
All Photos(1)

Key Documents

SML1193

Sigma-Aldrich

SU1498

≥98% (HPLC)

Synonym(s):

(2E)-2-Cyano-3-[4-hydroxy-3,5-bis(1-methylethyl)phenyl]-N-(3-phenylpropyl)-2-propenamide, (E)-N-(3-Phenylpropyl)-cyano-3,5-diisopropyl-4-hydroxycinnamamide, AG-1498, SU 1498, Tyrphostin SU 1498, Tyrphostin SU1498

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
966,00 kr.
25 MG
2.760,00 kr.

966,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
966,00 kr.
25 MG
2.760,00 kr.

About This Item

Empirical Formula (Hill Notation):
C25H30N2O2
CAS Number:
Molecular Weight:
390.52
UNSPSC Code:
12352200
NACRES:
NA.77

966,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

N(CCCc2ccccc2)C(=O)\C(=C\c1cc(c(c(c1)C(C)C)O)C(C)C)\C#N

InChI

1S/C25H30N2O2/c1-17(2)22-14-20(15-23(18(3)4)24(22)28)13-21(16-26)25(29)27-12-8-11-19-9-6-5-7-10-19/h5-7,9-10,13-15,17-18,28H,8,11-12H2,1-4H3,(H,27,29)/b21-13+

InChI key

JANPYFTYAGTSIN-FYJGNVAPSA-N

Related Categories

Biochem/physiol Actions

SU1498 is a potent and selective inhibitor of the VEGFR2 receptor kinase, Flk-1 with an IC50 value of 700 nM. SU1498 is a very weak inhibitor of PDGFR-kinase, EGFR-kinase and HER-2 kinase with IC50 values > 50 μM. SU1498 has been used to study the role of VEGFR2 (Flk-1) in a variety of biological activities including angiogenesis, tumor growth inhibition, and stem cell proliferation.
SU1498 is a potent and selective inhibitor of the VEGFR2 receptor kinase, Flk-1.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharavee Jaiprasart et al.
Oncotarget, 11(1), 99-114 (2020-02-01)
VEGF-mediated tumor angiogenesis is a validated clinical target in many cancers, but modest efficacy and rapid development of resistance are major challenges of VEGF-targeted therapies. To establish a molecular signature of this resistance in ovarian cancer, we developed preclinical tumor
Keisuke Shirakura et al.
EMBO molecular medicine, 15(4), e16128-e16128 (2023-02-07)
Vascular endothelial protein tyrosine phosphatase (VE-PTP) influences endothelial barrier function by regulating the activation of tyrosine kinase receptor Tie2. We determined whether this action is linked to the development of atherosclerosis by examining the influence of arterial shear stress on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service