Skip to Content
Merck
All Photos(1)

Key Documents

SML1136

Sigma-Aldrich

SW155246

≥98% (HPLC)

Synonym(s):

4-Chloro-N-(4-hydroxy-1-naphthalenyl)-3-nitro-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11ClN2O5S
CAS Number:
Molecular Weight:
378.79
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC=C(NS(=O)(C2=CC=C(Cl)C([N+]([O-])=O)=C2)=O)C3=C1C=CC=C3

InChI

1S/C16H11ClN2O5S/c17-13-6-5-10(9-15(13)19(21)22)25(23,24)18-14-7-8-16(20)12-4-2-1-3-11(12)14/h1-9,18,20H

InChI key

JDYDEJYIVZNWQU-UHFFFAOYSA-N

Biochem/physiol Actions

SW155246 is a potent and selective DNMT1 (DNA methyltransferase 1) inhibitor that demethylates cytosines in vivo in a cell-based assay without affecting protein levels or generating reactive oxygen species. SW155246 induces re-expression of the tumor suppressor geneRASSF1 in A549 cells.
SW155246 is a potent and selective DNMT1 (DNA methyltransferase 1) inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

light sensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service