Skip to Content
Merck
All Photos(1)

Documents

SML0727

Sigma-Aldrich

cisTPD

≥98% (HPLC)

Synonym(s):

(2a,4a,6a)-2,4,6-Triphenyl-1,3-dioxane, (4R,6S)-rel-2,4,6-Triphenyl-1,3-dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20O2
CAS Number:
Molecular Weight:
316.39
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

cisTPD is a constitutive androstane receptor (CAR) agonist that inhibits FOXO1 and HNF4 driven expression of the gluconeogeneis PEPCK and glucose-6-phosphatase. cisTPD reduces blood glucose levels, insulin sensitivity and weight gain in rats fed a high fat diet.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service