Skip to Content
Merck
All Photos(1)

Documents

SML0135

Sigma-Aldrich

GPV574

≥98% (HPLC)

Synonym(s):

1-[4-[3-[4-(2,3-dimethylphenyl)-1-piperazinyl]-2-hydroxypropoxy]phenyl]-ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N2O3
CAS Number:
Molecular Weight:
382.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥10 mg/mL (warmed to 60°C)

storage temp.

2-8°C

SMILES string

CC(=O)c1ccc(OCC(O)CN2CCN(CC2)c3cccc(C)c3C)cc1

InChI

1S/C23H30N2O3/c1-17-5-4-6-23(18(17)2)25-13-11-24(12-14-25)15-21(27)16-28-22-9-7-20(8-10-22)19(3)26/h4-10,21,27H,11-16H2,1-3H3

InChI key

KQZDGQRWGREOJP-UHFFFAOYSA-N

Biochem/physiol Actions

GPV574 is a derivative of the Class 1C antiarrhythmic drug propafenone. GPV574 inhibits the delayed rectifier K+ channel HERG with similar affinity and time constant as propafenone (IC50 = 5 mM), but unlike the parent compound, does not remain permanently trapped in the channel pore. After wash-out of GPV574, channel activity is recovered within 60 seconds.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service