Skip to Content
Merck
All Photos(1)

Key Documents

SML0051

Sigma-Aldrich

E-Resveratrol Trimethyl Ether

≥98% (HPLC)

Synonym(s):

1,3-dimethoxy-5-[(1E)-2-(4-methoxyphenyl)ethenyl]-benzene; Tri-O-methylresveratrol; Trimethoxy-trans-stilbene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18O3
CAS Number:
Molecular Weight:
270.32
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥34 mg/mL

shipped in

wet ice

storage temp.

−20°C

Application

E-Resveratrol Trimethyl Ether was used to study the effect of trans- and cis-stilbene polyphenols on mitochondrial clustering and p53-dependent apoptosis of cancer cells.

Biochem/physiol Actions

E-Resveratrol Trimethyl Ether inhibits the activation of SAPK/c-Jun N-terminal kinase (JNK), and reduces the levels of NF-κB and AP-1 in the nucleus. It acts as anti-migratory, anti-adhesive and anti-invasive agent.
E-Resveratrol Trimethyl Ether is a Resveratrol analog, SIRT activator
E-Resveratrol Trimethyl Ether, also known as trimethoxy-trans-stilbene, has been shown to inhibit migration and invasion of tumor cell lines such as HepG2 and A549 by a mechanism that involves the reduction of matrix metalloproteinase (MMP) expression.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Gosslau et al.
European journal of pharmacology, 587(1-3), 25-34 (2008-05-20)
We previously reported that 3,4,5,4'-tetramethoxy-trans-stilbene (MR-4) induces p53 and perinuclear mitochondrial clustering in cancer cells [Gosslau, A., Chen, M., Ho, C.-T., Chen, K.Y., 2005, A methoxy derivative of resveratrol analogue selectively induced activation of the mitochondrial apoptotic pathway in transformed
Ya-Ting Yang et al.
Molecular nutrition & food research, 53(3), 407-416 (2008-12-17)
Resveratrol (3,5,4'-trihydroxystilbene) is a natural polyphenol that presents various physiological activities. It has been reported that the methylated derivatives of resveratrol show better potential antifungal and antiproliferative activities than resveratrol. In the present study, we investigated the inhibitory effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service