Skip to Content
Merck
All Photos(1)

Documents

SMB00389

Sigma-Aldrich

Isoxanthohumol

≥99% (HPLC)

Synonym(s):

Isoxanthohumol, 4,7-Dihydroxy-5-methoxy-8-prenylflavanone, Humulol, IXN, 2,3-Dihydro-7-hydroxy-2-(4-hydroxyphenyl)-5-methoxy-8-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O5
CAS Number:
Molecular Weight:
354.40
Beilstein:
10199264
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

COc1cc(O)c(C\C=C(\C)C)c2OC(CC(=O)c12)c3ccc(O)cc3

InChI

1S/C21H22O5/c1-12(2)4-9-15-16(23)10-19(25-3)20-17(24)11-18(26-21(15)20)13-5-7-14(22)8-6-13/h4-8,10,18,22-23H,9,11H2,1-3H3

InChI key

YKGCBLWILMDSAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isoxanthohumol is a polyphenol found in Humulus lupulus.

Biochem/physiol Actions

Isoxanthohumol displays antioxidant, anti-inflammatory, and anti-angiogenic properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service