SIAL0639
Sigma® 25 cm cell culture flask
cap, vented, tissue-culture treated, sterile, pack of 10 × 20 ea
All Photos(1)
About This Item
Recommended Products
material
canted neck
polystyrene
sterility
sterile
feature
cap vented
graduations
packaging
pack of 10 × 20 ea
maximum volume
60 mL
surface area
25 cm2
working volume
5-7.5 mL
binding type
Tissue Culture (TC)-treated surface
General description
Available in a variety of sizes, designs and cap styles to meet your needs.
- Manufactured from optically clear virgin polystyrene
- Treated for optimal cell attachment
- Printed with lot numbers for ease in traceability
- 100% integrity tested
- Sterilized by gamma irradiation and certified nonpyrogenic
Legal Information
Sigma is a registered trademark of Sigma-Aldrich Co. LLC