Skip to Content
Merck
All Photos(1)

Documents

SCJ4600043

Sigma-Aldrich

CF 640R maleimide

suitable for fluorescence, ≥90% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Quality Level

Assay

≥90% (TLC)

form

solid

mol wt

~954 g/mol

solubility

DMF: soluble
DMSO: soluble
H2O: soluble

ε (extinction coefficient)

105000 at 642 nm

absorbance ratio

A280/642 nm (0.37)

fluorescence

λex 633, 635 or 640 nm (Flow cytometry laser line)
λex 633, 635 or 640 nm (Microscopy laser line)
λex 642 nm; λem 662 nm

suitability

suitable for fluorescence

shipped in

wet ice

storage temp.

−20°C

General description

CF640R is a rhodamine-based far-red fluorescent dye with excitation and emission maxima very similar to those of Cy®5 and Alexa Fluor 647. CF640R is much brighter than Cy®5 and at least as bright as Alexa Fluor 647. A major advantage of CF640R over Cy® 5 and Alexa Fluor 647 is its exceptional photostability. Cy®5 and Alexa Fluor 647 are cyanine-based dyes and, like other cyanine dyes in general, have intrinsically poor photostability. CF640R is also far brighter and more photostable than Atto 647N, another spectrally similar dye frequently used in single-molecule imaging. The combination of excellent brightness and photostability makes CF640R ideal for confocal microscopy, single- molecule imaging and other demanding applications based on fluorescence detection.

Features and Benefits

1. Has the best photostability among dyes with Cy®5-like spectra. 2. Yields highly fluorescent protien conjugates. 3. Very water-soluble and pH-insensitive.

Preparation Note

Protect from light

Reconstitution

Dissolve in DMSO

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: btinfo@biotium.com.
ALEXA FLUOR is a trademark of Life Technologies
Atto is a trademark of Atto-Tec GmbH
CF is a trademark of Biotium, Inc.
Cy is a registered trademark of Cytiva

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service