SBR00038
Hygromycin B
Min. 95% (HPLC ELSD), from Streptomyces hygroscopicus
Select a Size
381,00 kr.
Select a Size
About This Item
381,00 kr.
Recommended Products
Product Name
Hygromycin B, Min. 95% (HPLC ELSD), from Streptomyces hygroscopicus
biological source
Streptomyces hygroscopicus
Quality Level
form
solid
antibiotic activity spectrum
Gram-negative bacteria
Gram-positive bacteria
fungi
viruses
Mode of action
protein synthesis | interferes
storage temp.
2-8°C
SMILES string
N([C@@H]1[C@H]([C@@H]([C@H]([C@@H](C1)N)O)O[C@@H]2O[C@@H]([C@@H]([C@@H]3O[C@@]4(O[C@@H]32)O[C@@H]([C@@H]([C@@H]([C@H]4O)O)O)[C@H](N)CO)O)CO)O)C
InChI
1S/C20H37N3O13/c1-23-7-2-5(21)9(26)15(10(7)27)33-19-17-16(11(28)8(4-25)32-19)35-20(36-17)18(31)13(30)12(29)14(34-20)6(22)3-24/h5-19,23-31H,2-4,21-22H2,1H3/t5-,6-,7+,8-,9+,10-,11+,12-,13+,14-,15-,16+,17+,18-,19+,20+/m1/s1
InChI key
GRRNUXAQVGOGFE-XKIAHZFYSA-N
General description
Application
Biochem/physiol Actions
Antimicrobial Spectrum: Hygromycin B acts against bacteria, fungi and higher eukaryotic cells.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service