Skip to Content
Merck
All Photos(1)

Documents

SBPVR1

Sigma-Aldrich

SB PREDIVEZ Reagent Kit for MDR1/P-gp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

recombinant

expressed in human cells

form

liquid

shipped in

dry ice

storage temp.

−70°C

General description

SOLVO Biotechnology offers ready-to-use assay kits for efflux and uptake transporters. SOLVO′s PREDIVEZ Vesicular Transport Assay Kits are suitable for efflux transporter inhibition studies. The kits are designed to test the inhibitory effect of one or more test compounds on the vesicular transport of a probe substrate. In most cases a fluorescent probe substrate is included. Kits contain all reagents and solutions necessary to conduct the drug-transporter interaction assay of need.

Application

The PREDIVEZ Vesicular Transport Kit is a simple and powerful tool to investigate drug transporter interactions. It provides information on any interaction between the ABC transporter and the drug candidate that would affect the transport of the fluorescent or radiolabeled or cold.

PREDIVEZ vesicular transport kits contains buffers, probe substrate, and reference inhibitors needed to run vesicular transport assays (sufficient for 300 wells/kit –equivalent to existing 9-cpd PV kits). Assay protocol, calculation template, and example data file available on SOLVO website.

Transport membranes and control membranes are sold separately. This PREDIVEZ vesicular transport kit requires the control membrane SB K CTRL part number SBCT01-1EA and can be used with the below transporter membranes:

SB-MDR1-K-VT MDR1 human membrane vesicles part number SBVT01-1EA

Legal Information

PREDIVEZ is a trademark of Solvo Biotechnology

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service