Skip to Content
Merck
All Photos(1)

Key Documents

SAB5700609

Sigma-Aldrich

Anti-FasLG antibody produced in rabbit

Synonym(s):

ALPS1B, APT1LG1, APTL, CD178, CD95-L, CD95L, FASL, FASLG, Fas ligand, FasL, FasLG, TNFSF6, TNLG1A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
2.530,00 kr.

2.530,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
2.530,00 kr.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

2.530,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

14
31

species reactivity

human, mouse, rat

concentration

1.52 mg/ml

technique(s)

western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FasLG(356)

General description

This gene is a member of the tumor necrosis factor superfamily. The primary function of the encoded transmembrane protein is the induction of apoptosis triggered by binding to FAS. The FAS/FASLG signaling pathway is essential for immune system regulation, including activation-induced cell death (AICD) of T cells and cytotoxic T lymphocyte induced cell death. It has also been implicated in the progression of several cancers. Defects in this gene may be related to some cases of systemic lupus erythematosus (SLE). Alternatively spliced transcript variants have been described.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 100-200 of human FasLG (NP_000630.1).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service