Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502150

Sigma-Aldrich

Anti-ATP6V1H antibody produced in rabbit

affinity isolated antibody

Synonym(s):

V-ATPase 50/57 kDa subunits, V-ATPase subunit H, VMA13, Vacuolar proton pump subunit H, Vacuolar proton pump subunit SFD

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
2.420,00 kr.

2.420,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
2.420,00 kr.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

2.420,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP6V1H(51606)

General description

The gene ATP6V1H (ATPase H+ transporting V1 subunit H) is mapped to human chromosome 8q11.2. It encodes a lysosomal protein with a molar mass of 50/57kDa. This protein forms the subunit of vacuolar ATPase (V-ATPase), a protein widely found in various eukaryotic endomembrane organelles. V-ATPase is a member of the rotary ATPase family, the members of which use rotary motor mechanisms for the transportation of ions across membranes. V-ATPase has two domains, V0 and V1 that function in transportation of proton and hydrolysis of ATP, respectively. The V1 domain is composed of eight subunits, A-H.

Immunogen

The antiserum was produced against synthesized peptide derived from human ATP6V1H.

Immunogen Range: 341-390

Biochem/physiol Actions

The gene ATP6V1H (ATPase H+ transporting V1 subunit H) encodes the regulatory subunit H of the V1 domain of V-ATPase. This multisubunit complex of V-ATPase functions in the acidification of intracellular compartments via the energy generated from ATP hydrolysis. Therefore, V-ATPase participates in receptor-mediated endocytosis, intracellular trafficking processes and degradation of proteins. Down-regulation of ATP6V1H gene in human pancreatic islets has been associated with type 2 diabetes. Its expression is negatively associated with HbA1c levels. It has a positive regulatory effect on glucose-stimulated insulin secretion.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Decreased expression of ATP6V1H in type 2 diabetes: a pilot report on the diabetes risk study in Mexican Americans.
Molina MF
Biochemical and Biophysical Research Communications, 412, 728-731 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service