Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200293

Sigma-Aldrich

Anti-HAP1 (C-terminal) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-HAP2, Anti-HIP5, Anti-HLP, Anti-hHLP1, Anti-huntingtin-associated protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

human

concentration

~1.5 mg/mL

technique(s)

western blot: 1.5-3.0 μg/mL using lysates of A431 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... HAP1(9001)

General description

The gene HAP1 (huntingtin-associated protein 1) encodes a nuclear enzyme that is mainly expressed in neurons. It is the human homolog of exonuclease III protein from E. coli. The gene is mapped to human chromosome 17q21.2-q21.3.

Immunogen

synthetic peptide corresponding to a sequence located near the C-terminal of human HAP1, conjugated to KLH.

Biochem/physiol Actions

The gene HAP1 (huntingtin-associated protein 1) is a bifunctional enzyme that functions as a DNA repair enzyme as well as in redox activation of transcription factors. It binds to the Huntington′s disease (HD) protein huntingtin in a polyglutamine length-dependent manner. It associates with huntingtin via the cytoskeletal proteins dynactin and pericentriolar autoantigen protein 1 forming coiled-coils. It plays a role in vesicle trafficking by facilitating interactions among cytoskeletal, vesicular and motor proteins. It is an apurinic/apyrimidinic (AP) site-specific DNA repair endonuclease that removes the AP sites produced naturally or by cytotoxic drugs and radiation. It also has RNAse H activity and controls the redox state of certain proto-oncogene products, such as the transcription factor c-Jun. The HAP1 protein is a candidate for pathology associated with HD.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assignment of the huntingtin (Hdh) gene, the huntingtin associated protein (Hap1) gene, and the huntingtin interacting protein (Hip1) gene to rat chromosomes 14, 10, and 12 by radiation hybrid mapping and fluorescent in situ hybridization.
Zimdahl H
Cytogenetics and Cell Genetics, 94, 101-104 (2001)
A role for the human DNA repair enzyme HAP1 in cellular protection against DNA damaging agents and hypoxic stress.
Walker LJ
Nucleic Acids Research, 22, 4884-4889 (1994)
Huntingtin-associated protein 1 (HAP1) interacts with the p150Glued subunit of dynactin.
Engelender S
Human Molecular Genetics, 6, 2205-2212 (1997)
L J Walker et al.
Molecular and cellular biology, 13(9), 5370-5376 (1993-09-01)
The DNA binding activity of the c-jun proto-oncogene product is inhibited by oxidation of a specific cysteine residue (Cys-252) in the DNA binding domain. Jun protein inactivated by oxidation of this residue can be efficiently reactivated by a factor from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service