SAB3701453
Monoclonal Anti-Mesothelin antibody produced in mouse
clone MB-G10, purified immunoglobulin
Synonym(s):
Anti-CAK1 antigen, Anti-MB, Anti-MN, Anti-Mitotic arrest deficient 1 antibody, Anti-Pre-pro-megakaryocyte-potentiating factor
About This Item
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
MB-G10, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
ELISA: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG2a
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MSLN(10232)
Specificity
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service