Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501232

Sigma-Aldrich

Anti-gp120 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
2.960,00 kr.

2.960,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
2.960,00 kr.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

2.960,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

human immunodeficiency virus 1

concentration

1 mg/mL

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human immunodeficiency virus 1 ... env(155971)

Looking for similar products? Visit Product Comparison Guide

General description

gp120 Antibody: Human immunodeficiency virus type 1 (HIV-1) entry into target cells is directed by the envelope (Env) glycoproteins, which are present on the surface of HIV-1 virion or infected cells in the form of trimers consisting of gp120/gp41 complexes. The surface subunit, gp120, initiates the entry process by interacting sequentially with the CD4 receptor and a co-receptor CCR5 or CXCR4, thereby inducing a conformational change that allows the transmembrane (TM) gp41 subunit to mediate fusion between viral and target cell membranes. Cleavage of Env into its gp120 and gp41 components is necessary for activation of its fusogenic activity.

Immunogen

gp120 antibody was raised against 16 amino acid synthetic peptide representing the major HIV-1 consensus sequence near the N-terminal region of V1/V2 loop.The immunogen is located within amino acids 80 - 130 of gp120.

Application

Gp120 antibody can be used for detection of gp120 by Western blot at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service