Skip to Content
Merck
All Photos(1)

Key Documents

SAB2900872

Sigma-Aldrich

Anti-SLC37A2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Pp11662, SLC37A2, SPX2, Sugar phosphate exchanger 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

western blot: 1-2 μg/mL (Recombinant Protein)
western blot: 5 μg/mL (Cell Lysate)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Immunogen

Slc37A2 antibody was raised against a 15 amino acid peptide near the center of human Slc37A2.
TMEM184A antibody was raised against a 16 amino acid peptide near the carboxy terminus of human TMEM184A.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service