Skip to Content
Merck
All Photos(1)

Documents

SAB2501623

Sigma-Aldrich

Anti-SFRP1 (C-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FRP, Anti-FRP-1, Anti-FRP1, Secreted frizzled-related protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, bovine

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SFRP1(6422)

General description

The gene SFRP1 (secreted frizzled related protein 1) is mapped to human chromosome 8p22–p12, a region frequently deleted in breast cancer. It is highly expressed in normal breast tissue.

Immunogen

Peptide with sequence KNHECPTFQSVFK, from the C-terminal region of the protein sequence according to NP_003003.3

Biochem/physiol Actions

Secreted Frizzled-related protein 1 (SFRP1) is a secreted inhibitor of Wnt signaling pathway. DNA damage or oxidative stress causes cellular senescence, which leads to the overexpression of sfrp1. This protein induces senescence phenotypes via the inactivation of Wnt signaling and activation of the retinoblastoma (Rb) pathway. Mutations or aberrant methylation in this gene impair the induction of senescence in several cancers, including breast cancer, bladder cancer, and colorectal tumorigenesis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Veeck et al.
Oncogene, 25(24), 3479-3488 (2006-02-02)
The canonical Wnt signalling pathway plays a key role during embryogenesis and defects in this pathway have been implicated in the pathogenesis of various types of tumours, including breast cancer. The gene for secreted frizzled-related protein 1 (SFRP1) encodes a
Xiaobin Wang et al.
Oncology letters, 4(2), 334-338 (2012-07-31)
The aims of this study were to determine the methylation and expression status of secreted Frizzled-related protein 1 (SFRP1) in bladder cancer, to explore the mechanisms involved and to study the role of SFRP1 in the pathogenesis of bladder cancer.
Germaine M Caldwell et al.
Cancer research, 64(3), 883-888 (2004-02-12)
Regions of the short arm of chromosome 8 are deleted frequently in a range of solid tumors, indicating that tumor suppressor genes reside at these loci. In this study, we have examined the properties of the Wnt signaling antagonist secreted
David J Elzi et al.
Molecular and cellular biology, 32(21), 4388-4399 (2012-08-29)
Cellular senescence has emerged as a critical tumor suppressive mechanism in recent years, but relatively little is known about how senescence occurs. Here, we report that secreted Frizzled-related protein 1 (SFRP1), a secreted antagonist of Wnt signaling, is oversecreted upon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service