Skip to Content
Merck
All Photos(1)

Key Documents

SAB1407639

Sigma-Aldrich

Anti-ASXL2 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ASXH2, DKFZp686C1968, FLJ10898, KIAA1685

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
1
technique(s):
western blot: 1 μg/mL

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~98.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASXL2(55252)

General description

ASXL2 is a human homolog of the Drosophila asx gene. Drosophila asx is an enhancer of trithorax (see MIM 159555) and polycomb (see MIM 610231) (ETP) gene that encodes a chromatin protein with dual functions in transcriptional activation and silencing (Katoh and Katoh, 2003 [PubMed 12888926]).[supplied by OMIM

Immunogen

ASXL2 (AAH42999, 1 a.a. ~ 918 a.a) full-length human protein.

Sequence
MKRTKCADIDVETPDSILVNTNLRALINKHTFSVLPGDCQQRLLLLLPEVDRQVGPDGLMKLNGSALNNEFFTSAAQGWKERLSEGEFTPEMQVRIRQEIEKEKKVEPWKEQFFESYYGQSSGLSLEDSKKLTASPSDPKVKKTPAEQPKSMPVSEASLIRIVPVVSQSECKEEALQMSSPGRKEECESQGEVQPNFSTSSEPLLSSALNTHELSSILPIKCPKDEDLLEQKPVTSAEQESEKNHLTTASNYNKSESQESLVTSPSKPKSPGVEKPIVKPTAGAGPQETNMKEPLATLVDQSPESLKRKSSLTQEEAPVSWEKRPRVTENRQHQQPFQVSPQPFLNRGDRIQVRKVPPLKIPVSRISPMPFHPSQVSPRARFPVSITSPNRTGARTLADIKAKAQLVKAQRAAAAAAAAAAAAASVGGTIPGPGPGGGQGPGEGGEGQTARGGSPGSDRVSETGKGPTLELAGTGSRGGTRELLPCGPETQPQSETKTTPSQAQPHSVSGAQLQQTPPVPPTPAVSGACTSVPSPVHIEKLDNEKLNPTRATATVASVSHPQGPSSCRQEKAPSPTGFQLEDISTSQRFMLGFAGRRTSKPAMAGHYLLNISTYGRGSESFRRTHSVNPEDRFCLSSPTEALKMGYTDCKNATGESSSSKEDDTDEESTGDEQESVTVKEEPQVSQSAGKGDTSSGPHSRETLSTSDCLASKNVKAEIPLNEQTTLSKENYLFTRGQTFDEKTLARDLIQAAQKQMAHAVRGKAIRSSPELFSSTVLPLPADSPTHQPLLLPPLQTPKLYGSPTQIGPSYRGMINVSTSSDMDHNSAVPGSQVSSNVGDVMSFSVTVTTIPASQAMNPSSHGQTIPVQAFSEENSIEGTPSKCYCRLKAMIMCKGCGAFCHDDCIGPSKLCVSCLVVR

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela M Carter et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(31), 18401-18411 (2020-07-22)
Disparities in cancer patient responses have prompted widespread searches to identify differences in sensitive vs. nonsensitive populations and form the basis of personalized medicine. This customized approach is dependent upon the development of pathway-specific therapeutics in conjunction with biomarkers that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service