Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401170

Sigma-Aldrich

Anti-SERPIND1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

D22S673, HC2, HCF2, HCII, HLS2, LS2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
2.620,00 kr.

2.620,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
2.620,00 kr.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

2.620,00 kr.


Please contact Customer Service for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPIND1(3053)

General description

The product encoded by this gene is a serine proteinase inhibitor which rapidly inhibits thrombin in the presence of dermatan sulfate or heparin. The gene contains five exons and four introns. This protein shares homology with antithrombin III and other members of the alpha 1-antitrypsin superfamily. Mutations in this gene are associated with heparin cofactor II deficiency. (provided by RefSeq)

Immunogen

SERPIND1 (AAH35028.1, 1 a.a. ~ 499 a.a) full-length human protein.

Sequence
MKHSLNALLIFLIITSAWGGSKGPLDQLEKGGETAQSADPQWEQLNNKNLSMPLLPADFHKENTVTNDWIPEGEEDDDYLDLEKIFSEDDDYIDIVDSLSVSPTDSDVSAGNILQLFHGKSRIQRLNIVNAKFAFNLYRVLKDQVNTFDNIFIAPVGISTAMGMISLGLKGETHEQVHSILHFKDFVNASSKYEITTIHNLFRKLTHRLFRRNFGYTLRSVNDLYIQKQFPILLDFKTKVREYYFAEAQIADFSDPAFISKTNNHIMKLTKGLIKDALENIDPATQMMILNCIYFKGSWVNKFPVEMTHNHNFRLNEREVVKVSMMQTKGNFLAANDQELDCDILQLEYVGGISMLIVVPHKMSGMKTLEAQLTPGVVERWQKSMTNRTREVLLPKFKLEKNYNLVESLKLMGIRMLFDKNGNMAGISDQRIAIDLFKHQGTITVNEEGTQATTVTTVGFMPLSTQVRFTVDRPFLFLIYEHRTSCLLFMGRVANPSRS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service