S6507
Daminozide
Synonym(s):
Succinic acid mono(2,2-dimethylhydrazide)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO2CCH2CH2CONHN(CH3)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
1863230
EC Number:
MDL number:
UNSPSC Code:
12000000
Recommended Products
mp
162-164 °C (lit.)
storage temp.
2-8°C
SMILES string
CN(C)NC(=O)CCC(O)=O
InChI
1S/C6H12N2O3/c1-8(2)7-5(9)3-4-6(10)11/h3-4H2,1-2H3,(H,7,9)(H,10,11)
InChI key
NOQGZXFMHARMLW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service