Skip to Content
Merck
All Photos(1)

Key Documents

S0815

Sigma-Aldrich

Silyl[(6-O-tert-butyldimethyl)-2,3,-di-O-acetyl)-β-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C112H196O49Si7
Molecular Weight:
2523.32
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

>90%

form

powder

storage temp.

2-8°C

SMILES string

CC(=O)O[C@H]1[C@@H]2O[C@H](CO[Si](C)(C)C(C)(C)C)[C@@H](O[C@H]3O[C@H](CO)[C@@H](O[C@H]4O[C@H](CO)[C@@H](O[C@H]5O[C@H](CO)[C@@H](O[C@H]6O[C@H](CO)[C@@H](O[C@H]7O[C@H](CO)[C@@H](O[C@H]8O[C@H](CO)[C@@H](O2)[C@H](O)[C@H]8O)[C@H](O)[C@H]7O)[C@H](O)[C@H]6O)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)[C@@H]1OC(C)=O

InChI

1S/C52H88O37Si/c1-15(59)74-43-42-23(14-73-90(6,7)52(3,4)5)82-51(44(43)75-16(2)60)88-41-22(13-58)80-49(34(71)29(41)66)86-39-20(11-56)78-47(32(69)27(39)64)84-37-18(9-54)76-45(30(67)25(37)62)83-36-17(8-53)77-46(31(68)24(36)61)85-38-19(10-55)79-48(33(70)26(38)63)87-40-21(12-57)81-50(89-42)35(72)28(40)65/h17-51,53-58,61-72H,8-14H2,1-7H3/t17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43+,44-,45-,46-,47-,48-,49-,50-,51-/m1/s1

InChI key

NVNNUDVSLGUUHE-WDTBMEADSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service