Key Documents
RFA451811
Rotor for Eppendorf® 5418/5418R Centrifuges
FA-45-18-11, aerosol tight, to hold, 18 x 1.5/2.0 mL tubes, lid
About This Item
UNSPSC Code:
41103909
Product RFA451811 is not currently sold in your country. Contact Technical Service
feature
lid
to hold 18 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022652061 (NA)
Eppendorf® 5418707005 (EU)
parameter
16,873 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5418/5418R
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)