Skip to Content
Merck
All Photos(1)

Documents

R2636

Sigma-Aldrich

Rosamicin

from Micromonospora rosaria, ~98%

Synonym(s):

Juvenimicin A3, Rosaramicin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H51NO9
CAS Number:
Molecular Weight:
581.74
Beilstein:
1633003
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

biological source

Micromonospora rosaria

Assay

~98%

form

solid

storage temp.

2-8°C

SMILES string

[H]C(=O)C[C@H]1C[C@@H](C)C(=O)\C=C\[C@]2(C)O[C@@]2([H])[C@H](C)[C@@H](CC)OC(=O)C[C@@H](O)[C@H](C)[C@H]1O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C

InChI

1S/C31H51NO9/c1-9-25-20(5)29-31(6,41-29)12-10-23(34)17(2)14-21(11-13-33)28(19(4)24(35)16-26(36)39-25)40-30-27(37)22(32(7)8)15-18(3)38-30/h10,12-13,17-22,24-25,27-30,35,37H,9,11,14-16H2,1-8H3/b12-10+/t17-,18-,19+,20-,21+,22+,24-,25-,27-,28-,29+,30+,31+/m1/s1

InChI key

IUPCWCLVECYZRV-JZMZINANSA-N

Biochem/physiol Actions

Macrolide antibiotic similar to erythromycin, and with a similar spectrum of bactericidal activity. Against a wide variety of anaerobes, rosamicin is at least as potent as, and in some cases more potent than, erythromycin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yohei Iizaka et al.
Applied microbiology and biotechnology, 104(8), 3403-3415 (2020-02-28)
The cytochrome P450 monooxygenase RosC catalyzes the three-step oxidation reactions, which leads to the formation of a hydroxy, formyl, and carboxy group at C-20 during rosamicin biosynthesis in Micromonospora rosaria IFO13697. To determine if amino acid substitutions in RosC could
Yojiro Anzai et al.
Journal of industrial microbiology & biotechnology, 36(8), 1013-1021 (2009-05-02)
Some of the polyketide-derived bioactive compounds contain sugars attached to the aglycone core, and these sugars often impart specific biological activity to the molecule or enhance this activity. Mycinamicin II, a 16-member macrolide antibiotic produced by Micromonospora griseorubida A11725, contains
Isolation and characterization of 23-O-mycinosyl-20-dihydro-rosamicin: a new rosamicin analogue derived from engineered Micromonospora rosaria.
Yojiro Anzai et al.
The Journal of antibiotics, 63(6), 325-328 (2010-04-24)
A new Micromonospora-produced macrolide antibiotic, rosamicin.
G H Wagman et al.
The Journal of antibiotics, 25(11), 641-646 (1972-11-01)
K Funaishi et al.
The Journal of antibiotics, 43(8), 938-947 (1990-08-01)
Antibiotics 6108 A1, B, C and D, a new series of analogues of rosaramicin, were found together with rosaramicin, juvenimicin A4 and M-4365 A1 from the cultured broth of strain BA06108 which was assigned to be a new species of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service