Skip to Content
Merck
All Photos(1)

Key Documents

R2273

Sigma-Aldrich

D-Ribose-1-3H

≥95% (Radiochemical Purity, HPLC), aqueous ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥95% (Radiochemical Purity, HPLC)

form

aqueous ethanol solution

extent of labeling

10-20 Ci/mmol

shipped in

dry ice

storage temp.

−20°C

Packaging

Combi-vial

Physical form

Ethanol:water (9:1) solution

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service