R1031
RSK1, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
HU-1, MAPKAPK1A, RPS6KA1, S6K-alpha 1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
159-215 nmol/min·mg
mol wt
~108 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... RPS6KA1(6195)
General description
Ribosomal protein S6 kinase α-1 (RPS6KA1) or RSK1 is a member of the RSK (ribosomal S6 kinase) family that consists of growth factor-regulated serine/threonine kinases. RSK1 contains 2 nonidentical kinase catalytic domains. RSK1 encodes a predicted 735-amino acid protein containing 2 distinct consensus ATP-binding site sequences. RSK1 transcript is present in lymphocytes, skeletal muscle, liver, and adipose tissue.
Biochem/physiol Actions
Ribosomal S6 kinases (RSKs) are implicated in the activation of the mitogen-activated kinase (MAPK) cascade and the stimulation of cell proliferation and differentiation. The protein suppresses the pro-apoptotic function of BAD by phosphorylating it. RSK1 phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signaling pathway.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nucleic acids research, 48(16), 9361-9371 (2020-07-28)
Human Y-box binding protein 1 (YB-1) is a multifunctional protein and overexpressed in many types of cancer. It specifically recognizes DNA/RNA through a cold shock domain (CSD) and regulates nucleic acid metabolism. The C-terminal extension of CSD and the phosphorylation
Science (New York, N.Y.), 286(5443), 1365-1367 (1999-11-13)
Before fertilization, vertebrate eggs are arrested in metaphase of meiosis II by cytostatic factor (CSF), an activity that requires activation of the mitogen-activated protein kinase (MAPK) pathway. To investigate whether CSF arrest is mediated by the protein kinase p90Rsk, which
Rsk1 mediates a MEK-MAP kinase cell survival signal.
Current Biology (2000)
The American journal of physiology, 266(2 Pt 1), C351-C359 (1994-02-01)
Serine-threonine protein kinases in the ribosomal S6 kinase (rsk or p90rsk) family have been implicated as signaling intermediates in the cellular response to several growth factors. To investigate the molecular diversity of human p90rsk isoforms, mixed degenerate oligonucleotide polymerase chain
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service