Skip to Content
Merck
All Photos(1)

Key Documents

Q4972

Sigma-Aldrich

QS11 hydrate

≥98% (HPLC), powder

Synonym(s):

(2S)-2-[2-(Indan-5-yloxy)-9-(1,1′-biphenyl-4-yl)methyl)-9H-purin-6-ylamino]-3-phenyl-propan-1-ol hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H33N5O2 · xH2O
CAS Number:
Molecular Weight:
567.68 (anhydrous basis)
MDL number:
UNSPSC Code:
12352203

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

O.OC[C@H](Cc1ccccc1)Nc2nc(Oc3ccc4CCCc4c3)nc5n(Cc6ccc(cc6)-c7ccccc7)cnc25

Biochem/physiol Actions

QS11 is an inhibitor of GTPase activating protein of ADP-ribosylation factor 1 (ARFGAP1), as well as a synergizer of the Wnt/β-catenin signaling pathway.
QS11 is an inhibitor of GTPase activating protein of ADP-ribosylation factor 1 (ARFGAP1), as well as a synergizer of the Wnt/β-catenin signaling pathway. The Wnt/β-catenin signaling pathway regulates cell morphology, motility, proliferation, and differentiation during development. The GTPase activating protein of ADP-ribosylation factors (ARFGAPs) regulates the small GTPase ADP-ribosylation factors (ARFs) that mediate a number of biological processes, including vesicle trafficking and cytoskeleton reorganization. QS-11 binds and inhibits GTPase activating protein of ADP-ribosylation factor1 (ARFGAP1). Inhibition of ARFGAPs increases the level of activated ARF and subsequent β-catenin translocation leading to pathway activation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service