Skip to Content
Merck
All Photos(1)

Key Documents

PZ0345

Sigma-Aldrich

PF-06700841 tosylate salt

≥98% (HPLC)

Synonym(s):

[(1S)-2,2-Difluorocyclopropyl][3-[2-[(1-methyl-1H-pyrazol-4-yl)amino]-4-pyrimidinyl]-3,8-diazabicyclo[3.2.1]oct-8-yl]-methanone tosylate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
764,00 kr.

764,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
764,00 kr.

About This Item

Empirical Formula (Hill Notation):
C18H21F2N7O · C7H8O3S
CAS Number:
Molecular Weight:
561.60
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

764,00 kr.


Please contact Customer Service for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

FC1(F)C[C@H]1C(N2[C@H]3CN(C4=CC=NC(NC5=CN(C)N=C5)=N4)C[C@@H]2CC3)=O.CC6=CC=C(S(=O)(O)=O)C=C6

InChI key

FAKGOYNHHHOTEN-VDTKTRGNSA-N

Application

PF-06700841 tosylate salt has been used as an inhibitor of Janus kinase1 (JAK1) to study its therapeutic effect on the adjuvant induced arthritis (AIA) rat model.[1] It has also been used as an inhibitor of interferon-α/β receptor alpha chain (IFNAR1) signaling adaptor tyrosine kinase (2TYK2) in mice.[2]

Biochem/physiol Actions

PF-06700841 is an inhibitor of JAK1 and TYK2 kinases.
PF-06700841 is an inhibitor of JAK1 and TYK2 kinases. PF-06700841 has been investigated for the treatment of psoriasis and lupus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

308 Bone marrow/cord blood transplantation (BMCBT) ameliorates symptoms in some, but not all, subtypes of severe generalized junctional epidermolysis bullosa (JEB).
Hook K, et al.
The Journal of Investigative Dermatology, 137(5), S52-S52 (2017)
Andrew Fensome et al.
Journal of medicinal chemistry, 61(19), 8597-8612 (2018-08-17)
Cytokine signaling is an important characteristic of autoimmune diseases. Many pro-inflammatory cytokines signal through the Janus kinase (JAK)/Signal transducer and activator of transcription (STAT) pathway. JAK1 is important for the γ-common chain cytokines, interleukin (IL)-6, and type-I interferon (IFN) family
Christopher R Donnelly et al.
Nature, 591(7849), 275-280 (2021-01-15)
The innate immune regulator STING is a critical sensor of self- and pathogen-derived DNA. DNA sensing by STING leads to the induction of type-I interferons (IFN-I) and other cytokines, which promote immune-cell-mediated eradication of pathogens and neoplastic cells1,2. STING is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service