Skip to Content
Merck
All Photos(1)

Documents

PZ0137

Sigma-Aldrich

CP-471474

≥98% (HPLC)

Synonym(s):

2-[[[4-(4-Fluorophenoxy)phenyl]sulfonyl]amino]-N-hydrox y-2-methylpropanamide, PF-1626077

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17FN2O5S
CAS Number:
Molecular Weight:
368.38
MDL number:
UNSPSC Code:
41121801
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

CC(C)(NS(=O)(=O)c1ccc(Oc2ccc(F)cc2)cc1)C(=O)NO

InChI

1S/C16H17FN2O5S/c1-16(2,15(20)18-21)19-25(22,23)14-9-7-13(8-10-14)24-12-5-3-11(17)4-6-12/h3-10,19,21H,1-2H3,(H,18,20)

InChI key

QCOQJYRPDUMCNP-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

CP-471474 is a broad spectrum inhibitor of matrix metalloproteinases. CP-471474 has a low nM IC50 efficacy for MMP-2, MMP-3, MMP-9, and MMP-13. CP-471474 has low potency against MMP-1 (IC50 > 1 uM). CP-471474 prevents left ventricular remodeling after experimental myocardial infarction in mice. CP-471474 also inhibits cigarette smoke-induced lung inflammation and the progression of emphysema in guinea pig models.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service