Skip to Content
Merck
All Photos(1)

Documents

PCG3009

Sigma-Aldrich

TruPAGE LDS Sample Buffer

4 ×

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NB.22

concentration

4 ×

compatibility

for use with Sigma-Aldrich TruPAGE Precast Gels

storage temp.

2-8°C

General description

TruPAGE LDS Sample Buffer should ALWAYS be used to prepare protein samples for use with TruPAGE precast gels to achieve optimal band resolution and sharpness. The sample buffer is specifically formulated to complement the TruPAGE running buffer system and to deliver the optimal denaturing conditions without causing sample degradation. Effective protein denaturation occurs at 70 °C instead of the much higher temperature required with Laemmli buffer.

Application

TruPAGE LDS Sample Buffer-4× has been used in Western blotting.

Features and Benefits

TruPAGE LDS Sample Buffer is specially formulated to be used with TruPAGE Precast Gels to provide optimal sample loading conditions for gel electrophoresis. TruPAGE LDS Sample Buffer can also be supplemented with TruPAGE DTT Sample Reducer if reducing sample conditions are desired. Please refer to the gel migration chart to select the appropriate gel and running buffer combination for your experimental needs.

Legal Information

TruPAGE is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Registered report: Transcriptional amplification in tumor cells with elevated c-Myc.
Blum D, et al.
eLife, 4 (2015)
Registered report: Oncometabolite 2-hydroxyglutarate is a competitive inhibitor of α-ketoglutarate-dependent dioxygenases.
Evans B, et al.
eLife, 4 (2015)
Amanda L Wong et al.
Methods (San Diego, Calif.), 184, 102-111 (2020-02-06)
The development of universal, broadly applicable methods for histone extraction from animal cells and tissues has unlocked the ability to compare these epigenetic-influencing proteins across tissue types, healthy and diseased states, and cancerous versus normal cells. However, for plants and
Minzi Deng et al.
Experimental cell research, 395(1), 112175-112175 (2020-07-18)
Autophagy is a basic catabolic response that eukaryotic cells use to degrade unnecessary or dysfunctional cellular components in an orderly and regulated manner. It plays important roles in maintaining cellular homeostasis, energy homeostasis, response to environmental stimuli, and the development
Eleanor Wendy Trotter et al.
Open biology, 10(10), 200200-200200 (2020-10-15)
Each approach used to synchronize cell cycle progression of human cell lines presents a unique set of challenges. Induction synchrony with agents that transiently block progression through key cell cycle stages are popular, but change stoichiometries of cell cycle regulators

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service