Skip to Content
Merck
All Photos(1)

Key Documents

P8716

Sigma-Aldrich

Phalloidin, Biotin Labeled

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H83N13O13S4
Molecular Weight:
1286.61
MDL number:
UNSPSC Code:
12352200
eCl@ss:
34058011

Assay

≥90%

form

solid

solubility

DMSO/DMF: soluble
methanol: soluble

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Phalloidin is a cyclic peptide derived from Amanita phalloides. It is a filamentous-actin (F-actin) binding reagent. Phalloidin interacts with F-actin and blocks filament depolymerization. It is used for studying actin filament assembly.
Toxin that binds polymeric F actin, stabilizing it and interfering with the function of actin-rich structures.

Caution

Highly toxic

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparing lifeact and phalloidin for super-resolution imaging of actin in fixed cells.
Mazloom-Farsibaf, et al.
PLoS ONE, 16, e0246138-e0246138 (2021)
Influence of phalloidin on the formation of actin filament branches by Arp2/3 complex.
Mahaffy, et al.
Biochemistry, 47, 6460-6467 (2019)
Journal of the Chemical Society. Perkin Transactions 1, 2979-2979 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service