All Photos(1)
About This Item
Linear Formula:
CH3[CH2]1414COOH
CAS Number:
Molecular Weight:
256.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
ethanol solution
extent of labeling
40-60 mCi per mmol
mass shift
M+2
shipped in
dry ice
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCC[14C](O)=O
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service