Skip to Content
Merck
All Photos(2)

Key Documents

P6495

Sigma-Aldrich

Anti-PRMT6 (SE-15) antibody produced in rabbit

~0.9 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Protein Arginine Methyl Transferase 6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

~0.9 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using extracts of PRMT6 transfected 293T cells
indirect immunofluorescence: 3-4 μg/mL using paraformaldehyde fixed HeLa cells.
microarray: suitable
western blot: 0.25-0.5 μg/mL using cell extracts of PRMT6 transfected 293T cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRMT6(55170)

General description

Protein arginine methyltransferases (PRMTs) are divided in two major types, type I (including PRMT1, 3, 4, and 6) and type II (PRMT5). Protein arginine methyltransferase 6 (PRMT6), a histone H3R2 methyltransferase encodes protein arginine methyltransferase 6. This gene is located on human chromosome 1p13.3.

Specificity

Anti PRMT6 (SE-15) antibody reacts specifically with PRMT6.

Immunogen

synthetic peptide corresponding to amino acids 2-16 of human PRMT6 conjugated to KLH via a C-terminal added cysteine.

Application

Anti-PRMT6 (SE-15) antibody produced in rabbit may be used in:
  • western blotting
  • immunofluorescence
  • immunoprecipitation

Biochem/physiol Actions

Protein arginine methyltransferase 6 (PRMT6) enzyme is essential for the asymmetric dimethylation of H3R2 (H3R2me2a) especially at the arginine moiety. Overexpression of PRMT6 is seen in various human diseases including cancer. In MCF7 cells, deletion or blocking of this gene restores global DNA methylation. Absence of this gene results in the up-regulation of CDK-interacting protein 1 (p21 and p27) and cellular senescence. Antibodies specific for PRMT6 are an important tool for studying the biology of protein arginine methyltransferases (PRMTs).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For prolonged storage, freeze in working aliquots at −20 °C. Repeated freezing and thawing is not recom-mended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of phosphorylation and methylation on the function of the p16INK4a protein in non-small cell lung cancer A549 cells
Ma WL, et al.
Oncology Letters, 10(4), 2277-2282 (2015)
PRMT6 promotes endometrial cancer via AKT/mTOR signaling and indicates poor prognosis
Jiang N, et al.
The International Journal of Biochemistry & Cell Biology, 120(19), 105681-105681 (2020)
Significant association of PRMT 6 hypomethylation with colorectal cancer
Pan R, et al.
Journal of Clinical Laboratory Analysis, 32(9), e22590-e22590 (2018)
The arginine methyltransferase PRMT6 regulates DNA methylation and contributes to global DNA hypomethylation in cancer
Veland N, et al.
Cell Reports, 21(12), 3390-3397 (2017)
Structural basis of arginine asymmetrical dimethylation by PRMT6
Wu H, et al.
The Biochemical Journal, 473(19), 3049-3063 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service