P6134
2-Phenylethanol
≥98% (GC)
Synonym(s):
β-PEA, 2-Phenylethyl alcohol, Benzyl carbinol, PEA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH2CH2OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1905732
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
vapor density
4.21 (vs air)
vapor pressure
1 mmHg ( 58 °C)
Assay
≥98% (GC)
refractive index
n20/D 1.5317 (lit.)
bp
219-221 °C/750 mmHg (lit.)
mp
−27 °C (lit.)
density
1.020 g/mL at 20 °C (lit.)
SMILES string
OCCc1ccccc1
InChI
1S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2
InChI key
WRMNZCZEMHIOCP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
This product has been replaced by 77861-ALDRICH | 2-Phenylethanol ≥99.0% (GC)
recommended
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
215.6 °F - closed cup
Flash Point(C)
102 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Kunqiang Cui et al.
Biochemical and biophysical research communications, 435(2), 289-294 (2013-04-25)
Helicobacter pylori (H. pylori) is a major causative factor for gastrointestinal illnesses, H. pylori peptide deformylase (HpPDF) catalyzes the removal of formyl group from the N-terminus of nascent polypeptide chains, which is essential for H. pylori survival and is considered
Li-Zhen Wu et al.
Journal of pharmaceutical and biomedical analysis, 75, 55-63 (2013-01-15)
Ultra high-performance liquid chromatography coupled with electrospray ionization quadrupole time-of-flight tandem mass spectrometry (UHPLC/ESI-Q-TOF MS/MS) was used to investigate the MS fragmentation behaviors of flavone C-glycosides present in the extracts of five Trollius species. In this study, the primary MS
Ayako Tomizawa et al.
Oncology reports, 29(2), 425-429 (2012-12-12)
Caffeic acid esters have various biological activities, and we previously reported that undecyl caffeate (caffeic acid undecyl ester, CAUE), a new caffeic acid derivative, has strong pharmacological activity. The present study investigated the cytotoxicity of both CAUE and its parent
S Yanase et al.
Drug discoveries & therapeutics, 7(1), 29-35 (2013-03-26)
There is an increasing evidence that the oncogenic kinase PAK1 is responsible not only for malignant transformation, but also for several other diseases such as inflammatory diseases (asthma and arthritis), infectious diseases including malaria, AIDS, and flu, as well as
M Kamil Burgazli et al.
European review for medical and pharmacological sciences, 17(6), 720-728 (2013-04-24)
BACKGROUNG AND OBJECTIVES: Caffeic acid phenethyl ester (CAPE) is an active component of the resin propolis obtained from beehives. Propolis has a long history of medicinal use and a number of studies have already reported on some of its pharmaceutical
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service