Skip to Content
Merck
All Photos(1)

Key Documents

P5909

Sigma-Aldrich

Pentadecanolide

Synonym(s):

1-Oxa-2-cyclohexadecanone, 15-Hydroxypentadecanoic acid lactone, 15-Pentadecanolide, Pentalide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H28O2
CAS Number:
Molecular Weight:
240.38
Beilstein:
143710
EC Number:
MDL number:
UNSPSC Code:
12352211

bp

137 °C/2 mmHg (lit.)

mp

34-38 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCCCCCCCCCCO1

InChI

1S/C15H28O2/c16-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-17-15/h1-14H2

InChI key

FKUPPRZPSYCDRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Weeks et al.
Journal of applied toxicology : JAT, 40(10), 1421-1434 (2020-06-04)
In vitro metabolic stability of nine fragrance chemicals: p-tolyl acetate, cashmeran, ethylene brassylate, celestolide, galaxolide, traseolide, ambretone, tonalide and pentadecanolide, was evaluated in trout and human hepatocytes. The compounds were incubated with trout hepatocytes at 12°C and human hepatocytes at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service